Assay Details
Target Gene Details
Entrez Gene ID: | 60526 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
C2orf43, hLDAH |
Location: |
Chr.2:20682489-20823139 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LDAH |