Assay Details
Target Gene Details
Entrez Gene ID: | 2281 |
Gene Name: | FK506 binding protein 1B |
Gene Aliases: |
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase |
Location: |
Chr.2:24033205-24067743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP1B | XM_017003594.1 | XP_016859083.1 |
Target Gene Details
Entrez Gene ID: | 80304 |
Gene Name: | WD repeat and coiled coil containing |
Gene Aliases: |
C2orf44, PP384 |
Location: |
Chr.2:24029336-24047426 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WDCP | NM_001142319.1 | NP_001135791.1 | ||
NM_025203.2 | NP_079479.1 | |||
XM_017005029.1 | XP_016860518.1 | |||
AK025598.1 | BAB15186.1 | |||
BC035698.2 | AAH35698.1 | |||
DB292134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)