Assay Details
Target Gene Details
Entrez Gene ID: | 105374811 |
Gene Name: | uncharacterized LOC105374811 |
Gene Aliases: |
- |
Location: |
Chr.2:75146352-75279143 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105374811 |
Target Gene Details
Entrez Gene ID: | 6869 |
Gene Name: | tachykinin receptor 1 |
Gene Aliases: |
NK1R, NKIR, SPR, TAC1R |
Location: |
Chr.2:75046463-75199519 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TACR1 | NM_001058.3 | NP_001049.1 | ||
NM_015727.2 | NP_056542.1 | |||
AK289765.1 | ||||
AY462098.1 | AAR23925.1 | |||
BC074911.2 | AAH74911.1 | |||
BC074912.2 | AAH74912.1 | |||
BC095528.1 | AAH95528.1 | |||
M74290.1 | AAA60601.1 | |||
M76675.1 | AAA59936.1 | |||
M81797.1 | AAA59933.1 | |||
M84425.1 | AAA36641.1 | |||
M84426.1 | AAA36644.1 | |||
S62045.1 | AAB20168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv998145 | Chr.2:74969670 - 75241030 on Build GRCh38 | Loss | TACR1 POLE4 LOC105374811 MIR5000 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |