Assay Details
Target Gene Details
Entrez Gene ID: | 79623 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 14 |
Gene Aliases: |
GALNT15, GalNac-T10, GalNac-T14 |
Location: |
Chr.2:30888518-31138726 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GALNT14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2672526 | Chr.2:30901958 - 30909669 on Build GRCh38 | Deletion | GALNT14 |
esv3590210 | Chr.2:30901874 - 30909589 on Build GRCh38 | Loss | GALNT14 |
nsv833659 | Chr.2:30797706 - 30948639 on Build GRCh38 | Gain | GALNT14 CAPN13 |