Assay Details
Target Gene Details
Entrez Gene ID: | 51449 |
Gene Name: | prenylcysteine oxidase 1 |
Gene Aliases: |
PCL1 |
Location: |
Chr.2:70258099-70281185 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCYOX1 | NM_016297.3 | NP_057381.3 | ||
AB020715.1 | ||||
AF181490.1 | AAF16937.1 | |||
AK225302.1 | ||||
AK296478.1 | ||||
AK303043.1 | ||||
AK314453.1 | ||||
AK316013.1 | ||||
AY359063.1 | AAQ89422.1 | |||
BC007029.1 | AAH07029.1 | |||
BC033815.1 | AAH33815.1 | |||
BC051891.1 | AAH51891.1 | |||
DC417137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv961415 | Chr.2:70263316 - 70273124 on Build GRCh38 | Duplication |
![]() |
esv2667817 | Chr.2:70264390 - 70269399 on Build GRCh38 | Deletion |
![]() |
nsv518294 | Chr.2:70111212 - 70301469 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3591205 | Chr.2:70264550 - 70269576 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)