Assay Details
Target Gene Details
Entrez Gene ID: | 10018 |
Gene Name: | BCL2 like 11 |
Gene Aliases: |
BAM, BIM, BOD |
Location: |
Chr.2:111120914-111168445 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582699 | Chr.2:110763166 - 112172487 on Build GRCh38 | Loss | MIR4435-2 ANAPC1 MIR4771-2 TMEM87B MIR4435-2HG ACOXL LOC400997 PAFAH1B1P2 MERTK BCL2L11 FBLN7 |
nsv521956 | Chr.2:110634682 - 112341235 on Build GRCh38 | Loss | TMEM87B MIR4435-2HG LOC400997 BUB1 BCL2L11 FBLN7 MIR4435-2 ANAPC1 MIR4771-2 ACOXL PAFAH1B1P2 ZC3H8 MERTK SNORD132 ZC3H6 |
nsv834331 | Chr.2:111054562 - 111247101 on Build GRCh38 | Loss | MIR4435-2HG ACOXL LOC400997 BCL2L11 |
nsv428402 | Chr.2:111142206 - 111905020 on Build GRCh38 | Loss | MIR4435-2 ANAPC1 MIR4771-2 MIR4435-2HG PAFAH1B1P2 MERTK BCL2L11 |
dgv4047n100 | Chr.2:110308696 - 112358403 on Build GRCh38 | Loss | TMEM87B MIR4435-2HG LOC100288570 LOC400997 BUB1 BCL2L11 FBLN7 LOC105373553 MIR4435-2 LINC01106 ANAPC1 MIR4771-2 RGPD6 ACOXL LIMS4 PAFAH1B1P2 ZC3H8 MERTK SNORD132 ZC3H6 LOC105375809 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |