Assay Details
Target Gene Details
Entrez Gene ID: | 4664 |
Gene Name: | NGFI-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:190648860-190692766 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584252 | Chr.2:190654887 - 190674560 on Build GRCh38 | Loss | NAB1 |
nsv3082 | Chr.2:190627513 - 190688068 on Build GRCh38 | Insertion | NAB1 |
nsv834492 | Chr.2:190574138 - 190781895 on Build GRCh38 | Gain | NAB1 NEMP2 |
dgv1200e212 | Chr.2:190638342 - 190674560 on Build GRCh38 | Loss | NAB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |