Assay Details
Target Gene Details
Entrez Gene ID: | 101926913 |
Gene Name: | uncharacterized LOC101926913 |
Gene Aliases: |
- |
Location: |
Chr.2:170700368-170770766 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101926913 | NR_110185.1 | |||
BM931568.1 |