Assay Details
Target Gene Details
Entrez Gene ID: | 285148 |
Gene Name: | isoamyl acetate-hydrolyzing esterase 1 homolog |
Gene Aliases: |
- |
Location: |
Chr.2:9473638-9512403 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IAH1 | NM_001039613.2 | NP_001034702.1 | ||
NM_001320858.1 | NP_001307787.1 | |||
NM_001320859.1 | NP_001307788.1 | |||
NM_001320860.1 | NP_001307789.1 | |||
NM_001320863.1 | NP_001307792.1 | |||
XM_017003878.1 | XP_016859367.1 | |||
AK092982.1 | ||||
AK291984.1 | ||||
AK297648.1 | ||||
BC019061.1 | ||||
BC059410.1 | AAH59410.1 | |||
BC067865.1 | AAH67865.1 | |||
BC111025.1 | AAI11026.1 | |||
BF447213.1 | ||||
BM553525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv580915 | Chr.2:9168019 - 9704049 on Build GRCh38 | Gain | YWHAQ ITGB1BP1 ADAM17 CPSF3 LOC105373418 LOC101926982 IAH1 ASAP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |