Assay Details
Target Gene Details
Entrez Gene ID: | 6716 |
Gene Name: | steroid 5 alpha-reductase 2 |
Gene Aliases: |
- |
Location: |
Chr.2:31522486-31665651 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRD5A2 | NM_000348.3 | NP_000339.2 | ||
XM_011533069.2 | XP_011531371.1 | |||
XM_011533072.2 | XP_011531374.1 | |||
AK307235.1 | ||||
AK316597.1 | ||||
BC112252.1 | AAI12253.1 | |||
BC113641.1 | ||||
EF560740.1 | ||||
M74047.1 | AAA60586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3747n100 | Chr.2:31378589 - 31643808 on Build GRCh38 | Gain | XDH SRD5A2 |
esv3575212 | Chr.2:31372479 - 31643808 on Build GRCh38 | Gain | XDH SRD5A2 |
nsv581269 | Chr.2:31375957 - 31595187 on Build GRCh38 | Gain | XDH SRD5A2 |
esv3583710 | Chr.2:31525546 - 31532454 on Build GRCh38 | Loss | SRD5A2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |