Assay Details
Target Gene Details
Entrez Gene ID: | 90957 |
Gene Name: | DEAH-box helicase 57 |
Gene Aliases: |
DDX57 |
Location: |
Chr.2:38797729-38875883 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DHX57 | NM_198963.1 | NP_945314.1 | ||
XM_011533154.2 | XP_011531456.1 | |||
XM_011533155.2 | XP_011531457.1 | |||
XM_011533156.2 | XP_011531458.1 | |||
AB209249.1 | BAD92486.1 | |||
AF070590.1 | ||||
AF283512.1 | AAM73547.1 | |||
BC033636.1 | AAH33636.1 | |||
BC053623.1 | AAH53623.1 | |||
BC065278.1 | AAH65278.1 | |||
BC131534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss | LOC101929596 DHX57 GALM SRSF7 HNRNPLL CYP1B1-AS1 ATL2 GEMIN6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |