Assay Details
Target Gene Details
Entrez Gene ID: | 51072 |
Gene Name: | mediator of cell motility 1 |
Gene Aliases: |
C2orf4, CGI-27, MEMO, NS5ATP7 |
Location: |
Chr.2:31867188-32011052 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEMO1 | NM_001137602.2 | NP_001131074.1 | ||
NM_001301833.1 | NP_001288762.1 | |||
NM_001301852.1 | NP_001288781.1 | |||
NM_015955.3 | NP_057039.1 | |||
NR_126032.1 | ||||
NR_126034.1 | ||||
XM_011532892.2 | XP_011531194.1 | |||
XM_011532896.2 | XP_011531198.1 | |||
XM_017004255.1 | XP_016859744.1 | |||
AB041018.1 | BAD74066.1 | |||
AB041019.1 | BAD74067.1 | |||
AF132961.1 | AAD27736.1 | |||
AF363446.1 | AAL34462.1 | |||
AF529368.1 | AAQ09602.1 | |||
AK057760.1 | ||||
AK290753.1 | ||||
AK297128.1 | ||||
BC018733.1 | AAH18733.1 | |||
BC070046.1 | AAH70046.1 | |||
BC094681.1 | AAH94681.1 | |||
BG033287.1 | ||||
CD250945.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |