Assay Details
Target Gene Details
Entrez Gene ID: | 6646 |
Gene Name: | sterol O-acyltransferase 1 |
Gene Aliases: |
ACACT, ACAT, ACAT-1, ACAT1, SOAT, STAT |
Location: |
Chr.1:179293714-179358680 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOAT1 | NM_001252511.1 | NP_001239440.1 | ||
NM_001252512.1 | NP_001239441.1 | |||
NM_003101.5 | NP_003092.4 | |||
NR_045530.1 | ||||
XM_011509911.1 | XP_011508213.1 | |||
XM_011509912.1 | XP_011508214.1 | |||
AK026611.1 | ||||
AK290659.1 | ||||
AK294047.1 | ||||
AK300551.1 | ||||
AL833625.1 | ||||
BC028940.1 | AAH28940.1 | |||
DC344271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005278 | Chr.1:179228510 - 179402260 on Build GRCh38 | Gain | AXDND1 SOAT1 ABL2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |