Assay Details
Target Gene Details
Entrez Gene ID: | 1185 |
Gene Name: | chloride voltage-gated channel 6 |
Gene Aliases: |
CLC-6 |
Location: |
Chr.1:11806096-11843144 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLCN6 | NM_001256959.1 | NP_001243888.1 | ||
NM_001286.3 | NP_001277.1 | |||
NR_046428.1 | ||||
AK289999.1 | ||||
AK294764.1 | ||||
AK307098.1 | ||||
BC117420.1 | ||||
BC117424.1 | ||||
D28475.2 | ||||
DA229615.1 | ||||
DA935587.1 | ||||
X83378.1 | CAA58292.1 | |||
X96391.1 | CAA65255.1 | |||
X99473.1 | CAA67836.1 | |||
X99474.1 | CAA67837.1 | |||
X99475.1 | CAA67838.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)