Assay Details
Target Gene Details
Entrez Gene ID: | 79814 |
Gene Name: | agmatinase |
Gene Aliases: |
- |
Location: |
Chr.1:15571699-15585110 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGMAT | NM_024758.4 | NP_079034.3 | ||
AK130034.1 | ||||
AK225366.1 | ||||
AY057097.1 | AAL24446.1 | |||
BC005090.1 | AAH05090.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |