Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | FAM156A MAGEH1 MIR4536-1 TSR2 MIR6857 LOC100421746 MIR98 TSPYL2 SNORA11 FAM120C TRO PHF8 KDM5C PAGE2B USP51 PAGE5 ALAS2 FAM104B MIRLET7F2 WNK3 FOXR2 PFKFB1 MIR4536-2 HSD17B10 GNL3L MIR6895 GPR173 SMC1A PAGE2 KANTR FAM156B IQSEC2 MIR6894 MTRNR2L10 MAGED2 FGD1 HUWE1 PAGE3 SNORA109 APEX2 RIBC1 XAGE3 ITIH6 |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | FAM156A MAGEH1 MIR4536-1 TSR2 MIR6857 LOC100421746 MIR98 TSPYL2 SNORA11 FAM120C TRO PHF8 KDM5C PAGE2B USP51 PAGE5 ALAS2 FAM104B MIRLET7F2 WNK3 FOXR2 PFKFB1 MIR4536-2 HSD17B10 GNL3L MIR6895 GPR173 SMC1A PAGE2 KANTR FAM156B IQSEC2 MIR6894 MTRNR2L10 MAGED2 FGD1 HUWE1 PAGE3 SNORA109 APEX2 RIBC1 XAGE3 ITIH6 |