Assay Details
Target Gene Details
Entrez Gene ID: | 10134 |
Gene Name: | B-cell receptor-associated protein 31 |
Gene Aliases: |
6C6-AG, BAP31, CDM, DDCH, DXS1357E |
Location: |
Chr.X:153700492-153724746 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAP31 | NM_001139441.1 | NP_001132913.1 | ||
NM_001139457.2 | NP_001132929.1 | |||
NM_001256447.1 | NP_001243376.1 | |||
NM_005745.7 | NP_005736.3 | |||
AK057613.1 | ||||
AK125631.1 | ||||
AK222494.1 | BAD96214.1 | |||
AK223059.1 | BAD96779.1 | |||
AU132745.1 | ||||
BC014323.1 | AAH14323.1 | |||
BC065292.1 | AAH65292.1 | |||
BE729820.1 | ||||
BX474419.1 | ||||
CX164660.1 | ||||
X81109.1 | CAA57015.1 | |||
Z31696.1 | CAA83501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | OPN1MW3 RENBP FAM50A SLC6A8 AVPR2 LOC107987332 HAUS7 IKBKG ATP6AP1 BCAP31 HCFC1-AS1 ATP2B3 FAM58A TAZ PLXNA3 TMEM187 MECP2 EMD LOC105373386 OPN1MW2 MIR6858 FAM223A PNMA6A PDZD4 LOC105373377 ZNF275 FAM3A MIR3202-1 TEX28 ARHGAP4 CH17-340M24.3 GDI1 ZFP92 NAA10 LOC107983954 MAGEA1 LOC105373383 MIR718 PLXNB3 ABCD1 DNASE1L1 LAGE3 PNCK OPN1LW L1CAM LOC105373378 UBL4A MIR3202-2 IRAK1 SSR4 BGN TREX2 SNORA70 DUSP9 FLNA SLC10A3 HCFC1 CTAG1A LCA10 RPL10 G6PD TKTL1 LOC105373381 OPN1MW SRPK3 IDH3G |
esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss | BCAP31 SLC6A8 ABCD1 PNCK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |