Assay Details
Target Gene Details
Entrez Gene ID: | 56255 |
Gene Name: | thioredoxin related transmembrane protein 4 |
Gene Aliases: |
DJ971N18.2, PDIA14, TXNDC13 |
Location: |
Chr.20:7977353-8019829 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMX4 | NM_021156.3 | NP_066979.2 | ||
AK074838.1 | BAC11237.1 | |||
AK075404.1 | BAC11599.1 | |||
AK226005.1 | ||||
AL118494.1 | CAB56344.1 | |||
AY359004.1 | AAQ89363.1 | |||
BC033787.1 | AAH33787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059059 | Chr.20:7793987 - 8053708 on Build GRCh38 | Gain | LOC105372520 TMX4 HAO1 |
esv2422203 | Chr.20:7968779 - 8463601 on Build GRCh38 | Duplication | LOC105372520 PLCB1 TMX4 PLCB1-IT1 |
nsv585384 | Chr.20:7571617 - 8336371 on Build GRCh38 | Gain | LOC105372520 PLCB1 TMX4 PLCB1-IT1 HAO1 |
esv3893279 | Chr.20:7582024 - 7999743 on Build GRCh38 | Gain | TMX4 HAO1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |