Assay Details
Target Gene Details
Entrez Gene ID: | 5184 |
Gene Name: | peptidase D |
Gene Aliases: |
PROLIDASE |
Location: |
Chr.19:33386949-33521893 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PEPD | NM_000285.3 | NP_000276.2 | ||
NM_001166056.1 | NP_001159528.1 | |||
NM_001166057.1 | NP_001159529.1 | |||
AK225037.1 | ||||
AK290756.1 | ||||
AK290781.1 | ||||
AK291561.1 | ||||
AK293126.1 | ||||
AK294619.1 | ||||
BC004305.1 | AAH04305.1 | |||
BC015027.1 | AAH15027.1 | |||
BC028295.1 | AAH28295.1 | |||
BT006692.1 | AAP35338.1 | |||
CR541669.1 | CAG46470.1 | |||
J04605.1 | AAA60064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758755 | Chr.19:33286168 - 34090656 on Build GRCh38 | Gain | KCTD15 CHST8 CEBPA-AS1 CEBPA PEPD CEBPG LOC100996681 |
nsv522571 | Chr.19:33395448 - 33411903 on Build GRCh38 | Loss | PEPD |
nsv470138 | Chr.19:33387930 - 33418945 on Build GRCh38 | Gain | PEPD |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |