Assay Details
Target Gene Details
Entrez Gene ID: | 729767 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 18 |
Gene Aliases: |
- |
Location: |
Chr.19:51478543-51491261 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEACAM18 | NM_001278392.1 | NP_001265321.1 | ||
XM_011527258.2 | XP_011525560.1 |