Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | 10 | 3480 | NP_005348.2 |
XM_005258937.3 | 9 | 3247 | XP_005258994.1 | |
XM_005258938.4 | 10 | 2466 | XP_005258995.1 | |
XM_005258939.3 | 11 | 2504 | XP_005258996.2 | |
XM_005258940.3 | 10 | 2405 | XP_005258997.1 | |
XM_006723218.2 | 10 | 2605 | XP_006723281.1 | |
XM_017026810.1 | 10 | 2345 | XP_016882299.1 | |
AK292482.1 | ||||
BC070041.1 | 10 | 3480 | AAH70041.1 | |
U40002.1 | 10 | 3480 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 | ||||
DW455857.1 |
Target Gene Details
Entrez Gene ID: | 101930071 |
Gene Name: | uncharacterized LOC101930071 |
Gene Aliases: |
- |
Location: |
Chr.19:42397128-42408452 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101930071 | NR_126041.1 | |||
BU566442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LIPE-AS1 LIPE LOC101930071 CNFN |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | CNFN GSK3A ZNF574 RABAC1 MEGF8 LOC100505622 ARHGEF1 LOC390937 ERF PAFAH1B3 MIR6797 LIPE-AS1 POU2F2 MIR4323 ATP1A3 CD79A ZNF526 DEDD2 CIC GRIK5 TMEM145 LIPE LOC101930071 RPS19 LOC100505585 PRR19 MIR8077 |
nsv579632 | Chr.19:42400501 - 42408251 on Build GRCh38 | Gain | LIPE-AS1 LIPE LOC101930071 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | PAFAH1B3 LIPE-AS1 POU2F2 CNFN GSK3A ZNF526 CXCL17 CEACAM8 DEDD2 CIC TMEM145 LIPE MEGF8 LOC101930071 PRR19 ERF MIR8077 CEACAM1 |
dgv6460n54 | Chr.19:42400086 - 42402589 on Build GRCh38 | Loss | LIPE-AS1 LIPE LOC101930071 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | CNFN GSK3A ZNF574 RABAC1 MEGF8 LOC100505622 CEACAM4 ARHGEF1 CEACAM6 LOC390937 ERF PAFAH1B3 MIR6797 LIPE-AS1 POU2F2 CEACAM3 MIR4323 CEACAM5 ATP1A3 CD79A ZNF526 LYPD4 DMRTC2 DEDD2 CIC GRIK5 TMEM145 LIPE LOC101930071 RPS19 LOC100505585 CEACAM7 PRR19 MIR8077 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | PSG6 PSG3 PAFAH1B3 LIPE-AS1 PSG8 CNFN PSG11 PSG2 GSK3A CXCL17 CEACAM8 PSG10P CIC PSG1 LOC100289650 TMEM145 LIPE MEGF8 LOC101930071 PRR19 ERF MIR8077 PSG7 CEACAM1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |