Assay Details
Target Gene Details
Entrez Gene ID: | 4729 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit V2 |
Gene Aliases: |
CI-24k |
Location: |
Chr.18:9102623-9134345 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFV2 | NM_021074.4 | NP_066552.2 | ||
AK291865.1 | ||||
AK312129.1 | ||||
BC001632.1 | AAH01632.1 | |||
BC017487.1 | AAH17487.1 | |||
BC071689.1 | AAH71689.1 | |||
BG742948.1 | ||||
CB121843.1 | ||||
CR456928.1 | CAG33209.1 | |||
M22538.1 | AAA75390.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)