Assay Details
Target Gene Details
Entrez Gene ID: | 65264 |
Gene Name: | ubiquitin conjugating enzyme E2 Z |
Gene Aliases: |
HOYS7, USE1 |
Location: |
Chr.17:48908369-48929060 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2Z | NM_023079.4 | NP_075567.2 | ||
AB025426.1 | BAB87810.1 | |||
AK023917.1 | BAB14724.1 | |||
AK024030.1 | BAB14789.1 | |||
AK091644.1 | ||||
AK122770.1 | ||||
AK130303.1 | ||||
AK296865.1 | ||||
AL713782.1 | CAD28542.1 | |||
BC015169.1 | AAH15169.2 | |||
BC015890.1 | AAH15890.1 | |||
CR457322.1 | CAG33603.1 | |||
EF623992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain | LOC105371814 GIP UBE2Z SNF8 ATP5G1 IGF2BP1 |
nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss | CALCOCO2 LOC105371814 GIP UBE2Z SNF8 ATP5G1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |