Assay Details
Target Gene Details
Entrez Gene ID: | 79058 |
Gene Name: | ASPSCR1, UBX domain containing tether for SLC2A4 |
Gene Aliases: |
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9 |
Location: |
Chr.17:81977550-82018929 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASPSCR1 | NM_001251888.1 | NP_001238817.1 | ||
NM_024083.3 | NP_076988.1 | |||
NR_045351.1 | ||||
XM_011523601.2 | XP_011521903.1 | |||
XM_011523602.2 | XP_011521904.1 | |||
XM_017025036.1 | XP_016880525.1 | |||
XM_017025037.1 | XP_016880526.1 | |||
XM_017025038.1 | XP_016880527.1 | |||
XM_017025039.1 | XP_016880528.1 | |||
XM_017025040.1 | XP_016880529.1 | |||
XM_017025041.1 | XP_016880530.1 | |||
XM_017025042.1 | XP_016880531.1 | |||
XM_017025043.1 | XP_016880532.1 | |||
XM_017025044.1 | XP_016880533.1 | |||
AF324219.2 | AAK08959.2 | |||
AK057403.1 | BAB71472.1 | |||
AK057851.1 | BAB71595.1 | |||
AK226086.1 | ||||
AK290624.1 | ||||
BC006152.1 | AAH06152.1 | |||
BC018722.1 | AAH18722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952392 | Chr.17:81885825 - 82065724 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833569 | Chr.17:81838937 - 82011412 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)