Assay Details
Target Gene Details
Entrez Gene ID: | 23162 |
Gene Name: | mitogen-activated protein kinase 8 interacting protein 3 |
Gene Aliases: |
JIP-3, JIP3, JSAP1, SYD2, syd |
Location: |
Chr.16:1706183-1770317 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPK8IP3 | NM_001040439.1 | NP_001035529.1 | ||
NM_001318852.1 | NP_001305781.1 | |||
NM_015133.4 | NP_055948.2 | |||
XM_005255190.2 | XP_005255247.1 | |||
XM_006720869.2 | XP_006720932.1 | |||
XM_011522429.2 | XP_011520731.1 | |||
XM_011522430.2 | XP_011520732.1 | |||
XM_011522431.2 | XP_011520733.1 | |||
XM_011522432.2 | XP_011520734.1 | |||
XM_017023079.1 | XP_016878568.1 | |||
XM_017023080.1 | XP_016878569.1 | |||
XM_017023081.1 | XP_016878570.1 | |||
AB028989.2 | ||||
AB621811.1 | ||||
AK024437.1 | BAB15727.1 | |||
BC137123.1 | ||||
BC137124.1 | ||||
BC144486.1 | ||||
BC150266.1 | ||||
BY795428.2 | ||||
CN405915.1 | ||||
CR749696.1 | CAH18476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv571169 | Chr.16:1729694 - 1789022 on Build GRCh38 | Loss | MRPS34 MAPK8IP3 NUBP2 MIR3177 EME2 SPSB3 NME3 |
dgv4856n54 | Chr.16:1600945 - 1813519 on Build GRCh38 | Loss | MRPS34 CRAMP1 HN1L MIR3177 EME2 SPSB3 LOC105371046 MAPK8IP3 IFT140 NUBP2 NME3 HAGH IGFALS |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | PRSS27 MRPS34 PTX4 TELO2 SLC9A3R2 LOC105371046 SNHG9 C16orf91 POLR3K MIR4516 CHTF18 MIR662 FAHD1 AXIN1 LOC105371038 LINC00235 RAB40C HBA1 SNORA78 STUB1 GFER RAB26 CAPN15 TPSAB1 PGP PRR35 FBXL16 MIR5587 MIR3677 METTL26 TSR3 HN1L NARFL NOXO1 HS3ST6 ABCA3 MAPK8IP3 RHBDL1 MIR3176 TMEM8A GNPTG BRICD5 SNORD60 LMF1 DNASE1L2 WFIKKN1 NHLRC4 CRAMP1 LUC7L LOC652276 CCNF LOC100134368 TPSB2 KCTD5 MIR940 RGS11 MIR6859-4 MCRIP2 NUBP2 UBE2I RPS2 CEMP1 PDPK1 NTHL1 PRR25 TSC2 MSLN MIR6768 FAM173A EME2 HBQ1 LOC107984876 RPL23AP5 ECI1 RPL3L LMF1-AS1 SNRNP25 RPUSD1 MIR6767 NPRL3 LOC105371045 FLJ42627 SPSB3 SSTR5-AS1 C1QTNF8 MRPL28 MIR1225 WDR24 TMEM204 NTN3 MSRB1 RNPS1 RAB11FIP3 LOC105371047 LINC00254 TBC1D24 RNF151 CCDC154 PIGQ IGFALS MEIOB MPG LOC105371049 AMDHD2 HBZ SSTR5 HBA2 PDIA2 GNG13 CLCN7 MIR3180-5 HAGHL UNKL LOC105371184 CASKIN1 SYNGR3 METRN TPSD1 ERVK13-1 WASIR2 NME3 LOC729652 LOC101929280 JMJD8 NPW SNORA64 NME4 SNHG19 RHOT2 ATP6V0C TRAF7 PRSS29P NDUFB10 WDR90 IFT140 LOC100287175 DDX11L10 TPSG1 MIR4717 MIR6511B1 SNORA10 LOC106660606 MLST8 MIR3178 MIR3177 ABCA17P LOC107987233 C16orf59 HBM FAM234A CCDC78 SOX8 TBL3 DECR2 PKD1 E4F1 ZNF598 BAIAP3 HAGH RHBDF1 CACNA1H ARHGDIG |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss | MRPS34 MEIOB CRAMP1 TELO2 HN1L MIR3177 EME2 SPSB3 LOC105371046 MAPK8IP3 FAHD1 TMEM204 IFT140 NUBP2 LOC105371047 NME3 HAGH IGFALS |
nsv1160328 | Chr.16:1704619 - 1797867 on Build GRCh38 | Deletion | MRPS34 MAPK8IP3 NUBP2 MIR3177 EME2 SPSB3 NME3 IGFALS |
nsv518641 | Chr.16:1645775 - 1835631 on Build GRCh38 | Loss | MRPS34 MEIOB CRAMP1 HN1L MIR3177 EME2 SPSB3 MAPK8IP3 FAHD1 NUBP2 NME3 HAGH IGFALS |
nsv952900 | Chr.16:1724500 - 1827999 on Build GRCh38 | Deletion | MRPS34 MAPK8IP3 FAHD1 NUBP2 MIR3177 EME2 SPSB3 NME3 HAGH IGFALS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |