Assay Details
Target Gene Details
Entrez Gene ID: | 54620 |
Gene Name: | F-box and leucine rich repeat protein 19 |
Gene Aliases: |
CXXC11, Fbl19, JHDM1C |
Location: |
Chr.16:30923055-30948783 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FBXL19 | NM_001099784.2 | NP_001093254.2 | ||
NM_001282351.1 | NP_001269280.1 | |||
AK098777.1 | BAC05410.1 | |||
AK127701.1 | ||||
AK302669.1 | ||||
BC059173.1 | AAH59173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3638389 | Chr.16:30936363 - 30938698 on Build GRCh38 | Loss | FBXL19 |
nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss | PRSS36 ZNF668 ORAI3 BCKDK STX4 CTF1 MIR762 LOC101928762 FBXL19 KAT8 HSD3B7 MIR762HG BCL7C FBXL19-AS1 SETD1A VKORC1 PRSS53 FUS ZNF646 STX1B PRSS8 |
dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss | BCL7C ORAI3 FBXL19-AS1 STX4 SETD1A MIR4519 CTF1 MIR762 FBXL19 STX1B HSD3B7 MIR762HG |
nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion | BCL7C ORAI3 FBXL19-AS1 SETD1A CTF1 MIR762 FBXL19 STX1B HSD3B7 MIR762HG |
nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss | BCL7C ORAI3 FBXL19-AS1 MIR4519 CTF1 MIR762 FBXL19 MIR762HG |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |