Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | IL27 APOBR SULT1A1 SGF29 CLN3 SULT1A2 NUPR1 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | IL27 LOC101928215 XPO6 ATP2A1-AS1 SULT1A1 CLN3 MIR4721 RABEP2 EIF3CL LAT SBK1 SULT1A2 APOBR TUFM MIR6862-2 NPIPB8 NFATC2IP EIF3C NUPR1 NPIP ATP2A1 MIR4517 SPNS1 ATXN2L CD19 NPIPB6 NPIPB9 MIR6862-1 SGF29 RRN3P2 SH2B1 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | IL27 LOC101928215 ATP2A1-AS1 SULT1A1 CLN3 MIR4721 RABEP2 EIF3CL LAT SULT1A2 LOC107984832 APOBR TUFM MIR6862-2 NPIPB8 NFATC2IP EIF3C NUPR1 NPIP ATP2A1 MIR4517 SPNS1 ATXN2L CD19 NPIPB9 MIR6862-1 SGF29 RRN3P2 SH2B1 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | IL27 APOBR EIF3CL NPIPB6 MIR6862-1 SGF29 CLN3 NUPR1 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | APOBR CLN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |