Assay Details
Target Gene Details
Entrez Gene ID: | 9727 |
Gene Name: | RAB11 family interacting protein 3 |
Gene Aliases: |
CART1, Rab11-FIP3 |
Location: |
Chr.16:425621-522485 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB11FIP3 | NM_014700.3 | NP_055515.1 | ||
XM_005255713.3 | XP_005255770.1 | |||
XM_005255717.3 | XP_005255774.1 | |||
XM_005255718.3 | XP_005255775.1 | |||
XM_017023907.1 | XP_016879396.1 | |||
AB014565.1 | ||||
AF395731.1 | AAL12940.1 | |||
BC051360.1 | AAH51360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | TBL3 CLCN7 LOC105371038 SOX8 LOC105371047 RPUSD1 UNKL CCNF SPSB3 RNF151 RPS2 AXIN1 ABCA3 RGS11 WASIR2 MIR3178 LMF1-AS1 MEIOB ATP6V0C CAPN15 MIR4717 MRPL28 C16orf91 RAB26 KCTD5 MIR662 NUBP2 TBC1D24 LOC100134368 RHBDF1 BRICD5 FAHD1 MIR6511B1 JMJD8 MIR6859-4 SNHG19 DDX11L10 NARFL GNG13 CEMP1 LOC105371184 LOC652276 RHOT2 SNORA64 E4F1 SLC9A3R2 LOC101929280 FBXL16 STUB1 HAGH AMDHD2 ECI1 NTN3 PTX4 C1QTNF8 HBQ1 MIR3677 TSR3 MIR940 SNORA10 ZNF598 LMF1 METRN FAM173A MSLN HBA1 RPL3L ARHGDIG GFER WDR90 TPSB2 PDPK1 HBM LOC729652 LINC00235 MCRIP2 LOC105371049 PRSS27 PKD1 SNORA78 LOC105371045 MIR6768 IFT140 SSTR5 TPSG1 PDIA2 HN1L LOC106660606 MIR4516 LOC107984876 NHLRC4 LUC7L NPRL3 ERVK13-1 HS3ST6 SNORD60 EME2 SSTR5-AS1 RAB40C MSRB1 PRSS29P MPG ABCA17P MIR6767 HAGHL WDR24 IGFALS MAPK8IP3 RHBDL1 MIR3176 LINC00254 LOC105371046 FAM234A CCDC78 GNPTG PRR25 CRAMP1 NTHL1 CCDC154 CASKIN1 SNHG9 NPW TPSD1 PGP UBE2I NDUFB10 MIR3177 RPL23AP5 DECR2 MRPS34 FLJ42627 DNASE1L2 LOC107987233 POLR3K RNPS1 CACNA1H HBZ MLST8 METTL26 TMEM8A TELO2 PIGQ NME3 HBA2 LOC100287175 PRR35 CHTF18 SNRNP25 TPSAB1 TRAF7 TMEM204 RAB11FIP3 C16orf59 TSC2 NME4 MIR5587 MIR1225 BAIAP3 WFIKKN1 SYNGR3 NOXO1 MIR3180-5 |
esv3637576 | Chr.16:439034 - 480097 on Build GRCh38 | Gain | RAB11FIP3 |
esv3637563 | Chr.16:263248 - 505590 on Build GRCh38 | Gain | NME4 RPL23AP5 TMEM8A DECR2 AXIN1 RGS11 MRPL28 PDIA2 ARHGDIG FAM234A LOC100134368 RAB11FIP3 |
nsv526903 | Chr.16:451362 - 810673 on Build GRCh38 | Loss | JMJD8 LINC00235 MCRIP2 LOC105371038 RPUSD1 NARFL GNG13 LOC105371184 RHOT2 LOC101929280 FBXL16 LOC107987233 STUB1 METTL26 NHLRC4 PIGQ CAPN15 LOC100287175 PRR35 CHTF18 RAB40C HAGHL RAB11FIP3 MIR662 WDR24 METRN MIR5587 FAM173A MSLN RHBDL1 MIR3176 WFIKKN1 CCDC78 WDR90 PRR25 |
nsv1700 | Chr.16:457843 - 476640 on Build GRCh38 | Insertion | RAB11FIP3 |
nsv570954 | Chr.16:342461 - 530124 on Build GRCh38 | Loss | NME4 RPL23AP5 TMEM8A DECR2 AXIN1 LINC00235 CAPN15 MRPL28 LOC100134368 RAB11FIP3 |
dgv32n68 | Chr.16:245507 - 476226 on Build GRCh38 | Loss | NME4 RPL23AP5 TMEM8A DECR2 AXIN1 RGS11 MRPL28 PDIA2 ARHGDIG FAM234A LOC100134368 RAB11FIP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |