Assay Details
Target Gene Details
Entrez Gene ID: | 1201 |
Gene Name: | CLN3, battenin |
Gene Aliases: |
BTS, JNCL |
Location: |
Chr.16:28466653-28492302 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SULT1A1 SGF29 APOBR IL27 SULT1A2 NUPR1 CLN3 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | LOC101928215 SULT1A1 NPIPB8 XPO6 NPIP CD19 SPNS1 EIF3C MIR6862-2 APOBR EIF3CL MIR4517 NPIPB9 NUPR1 RABEP2 NPIPB6 RRN3P2 SGF29 IL27 LAT SULT1A2 SBK1 ATXN2L MIR6862-1 ATP2A1-AS1 SH2B1 MIR4721 ATP2A1 TUFM NFATC2IP CLN3 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | LOC101928215 SULT1A1 NPIPB8 NPIP CD19 SPNS1 EIF3C MIR6862-2 APOBR EIF3CL MIR4517 NPIPB9 NUPR1 RABEP2 RRN3P2 SGF29 IL27 LAT SULT1A2 ATXN2L MIR6862-1 ATP2A1-AS1 SH2B1 MIR4721 ATP2A1 TUFM NFATC2IP LOC107984832 CLN3 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | NPIPB6 MIR6862-1 SGF29 APOBR IL27 EIF3CL NUPR1 CLN3 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | APOBR CLN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |