Assay Details
Target Gene Details
Entrez Gene ID: | 1854 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
dUTPase |
Location: |
Chr.15:48331018-48343373 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DUT | NM_001025248.1 | NP_001020419.1 | ||
NM_001025249.1 | NP_001020420.1 | |||
NM_001948.3 | NP_001939.1 | |||
XM_011521315.2 | XP_011519617.1 | |||
AB049113.2 | BAB13724.1 | |||
AK000629.1 | ||||
AK291515.1 | ||||
AK298464.1 | ||||
AK312122.1 | ||||
BC033645.1 | AAH33645.1 | |||
BC070339.1 | AAH70339.1 | |||
BC110377.1 | ||||
BG505144.1 | ||||
CR541720.1 | CAG46521.1 | |||
CR541781.1 | CAG46580.1 | |||
L11877.1 | AAA36801.1 | |||
M89913.1 | AAA58444.1 | |||
U31930.1 | AAC50418.1 | |||
U62891.1 | AAC51123.1 | |||
U90223.1 | AAB94642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3636412 | Chr.15:48322023 - 48347079 on Build GRCh38 | Gain | DUT |
esv2760030 | Chr.15:48216841 - 48371581 on Build GRCh38 | Loss | SLC12A1 DUT |
esv3569565 | Chr.15:48317122 - 48346517 on Build GRCh38 | Gain | DUT |
nsv1053885 | Chr.15:48314149 - 48346517 on Build GRCh38 | Gain | DUT |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |