Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | C14orf79 GPR132 AHNAK2 CDCA4 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | LOC102723354 C14orf79 GPR132 JAG2 AHNAK2 CDCA4 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 PLD4 AHNAK2 CDCA4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | RD3L LINC00638 SIVA1 BAG5 SNORA28 ASPG LOC105378183 KIF26A CKB KLC1 BRF1 PACS2 BTBD6 EIF5 LOC105370687 LOC388022 NUDT14 GPR132 LOC107984648 TDRD9 INF2 JAG2 APOPT1 C14orf180 PPP1R13B MIR4710 TRMT61A AHNAK2 CEP170B MIR203B C14orf2 CDCA4 MIR203A LOC101929634 ZBTB42 MARK3 AKT1 PLD4 MIR6765 TMEM179 LOC105370691 LOC102723354 C14orf79 ZFYVE21 LINC00637 XRCC3 ADSSL1 LOC105370708 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | LOC102723354 C14orf79 GPR132 JAG2 BRF1 MIR6765 AHNAK2 BTBD6 NUDT14 CDCA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |