Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | AHNAK2 C14orf79 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | CDCA4 AHNAK2 GPR132 C14orf79 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | JAG2 LOC102723354 CDCA4 AHNAK2 GPR132 C14orf79 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | CDCA4 AHNAK2 C14orf79 PLD4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | AHNAK2 C14orf79 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | KLC1 LOC102723354 MARK3 LOC388022 ASPG CDCA4 EIF5 MIR203A LOC105370687 BTBD6 TRMT61A C14orf180 KIF26A JAG2 MIR6765 LINC00638 ADSSL1 BRF1 RD3L C14orf79 ZFYVE21 LOC101929634 XRCC3 AHNAK2 INF2 GPR132 SIVA1 LOC105370708 NUDT14 LOC107984648 APOPT1 CEP170B PACS2 LOC105370691 BAG5 CKB MIR4710 ZBTB42 TMEM179 SNORA28 PPP1R13B AKT1 LINC00637 PLD4 MIR203B C14orf2 LOC105378183 TDRD9 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | JAG2 LOC102723354 MIR6765 CDCA4 AHNAK2 BRF1 GPR132 C14orf79 BTBD6 NUDT14 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | JAG2 LOC102723354 MIR6765 CDCA4 AHNAK2 BRF1 GPR132 C14orf79 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |