Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 | ||
XM_005267299.1 | XP_005267356.1 | |||
AK094143.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | AHNAK2 C14orf79 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | AHNAK2 GPR132 C14orf79 CDCA4 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | AHNAK2 JAG2 GPR132 LOC102723354 C14orf79 CDCA4 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | AHNAK2 C14orf79 PLD4 CDCA4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | AHNAK2 C14orf79 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | C14orf2 BAG5 ZFYVE21 LOC102723354 C14orf79 NUDT14 XRCC3 CEP170B MIR4710 INF2 APOPT1 EIF5 TMEM179 MIR203B LOC101929634 BTBD6 PPP1R13B SNORA28 LOC105378183 C14orf180 AKT1 CKB MIR203A AHNAK2 PACS2 ADSSL1 TDRD9 BRF1 LINC00638 PLD4 LOC388022 LINC00637 MARK3 KLC1 MIR6765 CDCA4 ZBTB42 RD3L KIF26A LOC105370708 SIVA1 JAG2 LOC105370687 LOC107984648 TRMT61A ASPG GPR132 LOC105370691 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | AHNAK2 JAG2 BRF1 GPR132 LOC102723354 C14orf79 BTBD6 NUDT14 MIR6765 CDCA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |