Assay Details
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLEU2 | NR_002612.1 | |||
AY455757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580637 | Chr.13:50006792 - 50829882 on Build GRCh38 | Loss | DLEU2 DLEU7 MIR15A MIR16-1 ST13P4 KCNRG DLEU1-AS1 DLEU1 TRIM13 DLEU7-AS1 |
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss | DLEU2 RNASEH2B KCNRG DLEU1-AS1 TRIM13 MIR3613 DLEU7 MIR15A RNASEH2B-AS1 MIR16-1 INTS6 GUCY1B2 SERPINE3 ST13P4 SPRYD7 MIR5693 INTS6-AS1 FAM124A DLEU1 DLEU7-AS1 LINC00371 |
nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss | DLEU2 MIR15A MIR16-1 ST13P4 KCNRG DLEU1-AS1 DLEU1 TRIM13 MIR3613 |