Assay Details
Target Gene Details
Entrez Gene ID: | 28992 |
Gene Name: | MACRO domain containing 1 |
Gene Aliases: |
LRP16 |
Location: |
Chr.11:63998554-64166652 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MACROD1 | NM_014067.3 | NP_054786.2 | ||
XM_005273939.4 | XP_005273996.1 | |||
XM_011544970.2 | XP_011543272.1 | |||
AF086206.1 | ||||
AF202922.2 | AAF15294.2 | |||
BC000270.1 | AAH00270.2 | |||
BC003188.1 | AAH03188.1 | |||
BC007297.1 | AAH07297.1 | |||
BC008316.1 | AAH08316.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | MIR7155 BAD LOC105369340 FERMT3 FKBP2 MACROD1 PPP1R14B GPR137 CCDC88B VEGFB KCNK4-TEX40 TEX40 OTUB1 DNAJC4 FLRT1 COX8A NUDT22 TRPT1 PLCB3 NAA40 STIP1 PRDX5 ESRRA TRMT112 KCNK4 |
nsv1042075 | Chr.11:63964982 - 64045692 on Build GRCh38 | Loss | OTUB1 MACROD1 FLRT1 COX8A |
esv29993 | Chr.11:63971170 - 64096739 on Build GRCh38 | Loss | OTUB1 MACROD1 FLRT1 COX8A |
nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss | RCOR2 OTUB1 MARK2 MACROD1 COX8A NAA40 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |