Assay Details
Target Gene Details
Entrez Gene ID: | 54843 |
Gene Name: | synaptotagmin like 2 |
Gene Aliases: |
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A |
Location: |
Chr.11:85694221-85811182 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832222 | Chr.11:85694669 - 85892938 on Build GRCh38 | Gain | SYTL2 CCDC83 |
nsv468768 | Chr.11:85684137 - 85725309 on Build GRCh38 | Loss | CCDC89 SYTL2 |
esv2759845 | Chr.11:85469436 - 85892945 on Build GRCh38 | Gain+Loss | TMEM126A CCDC89 CREBZF TMEM126B DLG2 SYTL2 CCDC83 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |