Assay Details
Target Gene Details
Entrez Gene ID: | 114971 |
Gene Name: | protein tyrosine phosphatase, mitochondrial 1 |
Gene Aliases: |
DUSP23, MOSP, PLIP, PNAS-129 |
Location: |
Chr.11:47565430-47573461 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPMT1 | NM_001143984.1 | NP_001137456.1 | ||
NM_175732.2 | NP_783859.1 | |||
AF277187.1 | AAK07545.1 | |||
AK294640.1 | ||||
AY333987.1 | AAP94732.1 | |||
BC014048.1 | AAH14048.1 | |||
BC020242.1 | AAH20242.1 | |||
DA476984.1 | ||||
DR004030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832142 | Chr.11:47496403 - 47659758 on Build GRCh38 | Loss | FAM180B NDUFS3 PTPMT1 C1QTNF4 KBTBD4 AGBL2 MTCH2 CELF1 |
nsv825872 | Chr.11:47543873 - 47638778 on Build GRCh38 | Gain | FAM180B NDUFS3 PTPMT1 C1QTNF4 KBTBD4 MTCH2 CELF1 |
nsv314 | Chr.11:47540012 - 47584704 on Build GRCh38 | Deletion | NDUFS3 PTPMT1 KBTBD4 CELF1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |