Assay Details
Target Gene Details
Entrez Gene ID: | 26019 |
Gene Name: | UPF2 regulator of nonsense transcripts homolog (yeast) |
Gene Aliases: |
HUPF2, RENT2, smg-3 |
Location: |
Chr.10:11920022-12043170 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UPF2 | NM_015542.3 | NP_056357.1 | ||
NM_080599.2 | NP_542166.1 | |||
XM_017016034.1 | XP_016871523.1 | |||
AB037829.1 | ||||
AF301013.1 | AAG33225.1 | |||
AF318574.1 | AAG60689.1 | |||
AK000764.1 | BAA91369.1 | |||
AK074375.1 | BAB85064.1 | |||
AK096191.1 | BAC04721.1 | |||
AK295209.1 | ||||
AL080198.1 | CAB45771.1 | |||
AY013249.1 | AAG48509.1 | |||
BC114964.1 | ||||
BC115737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831785 | Chr.10:11928851 - 12124076 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)