Assay Details
Target Gene Details
Entrez Gene ID: | 55326 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 5 |
Gene Aliases: |
1AGPAT5, LPAATE |
Location: |
Chr.8:6708357-6761503 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGPAT5 | NM_018361.3 | NP_060831.2 | ||
XM_011534751.2 | XP_011533053.1 | |||
AF375789.1 | AAK54809.1 | |||
AK002072.1 | BAA92069.1 | |||
AK310545.1 | ||||
AL136587.1 | CAB66522.1 | |||
BC023550.2 | AAH23550.1 | |||
BC068519.1 | AAH68519.1 | |||
BC080537.1 | AAH80537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1021684 | Chr.8:6492570 - 6951933 on Build GRCh38 | Loss | MIR4659A MCPH1 DEFA6 AGPAT5 DEFB1 MIR8055 GS1-24F4.2 MIR4659B MCPH1-AS1 DEFA8P ANGPT2 XKR5 DEFA4 |
nsv610004 | Chr.8:6544845 - 6915391 on Build GRCh38 | Gain | MIR4659A MCPH1 XKR5 AGPAT5 DEFB1 MIR8055 GS1-24F4.2 MIR4659B MCPH1-AS1 ANGPT2 |
nsv1025976 | Chr.8:6615427 - 6829450 on Build GRCh38 | Loss | MIR4659A MCPH1 XKR5 AGPAT5 MIR8055 MIR4659B MCPH1-AS1 |
nsv6059 | Chr.8:6693878 - 6726740 on Build GRCh38 | Insertion | AGPAT5 MCPH1-AS1 |
esv2422383 | Chr.8:6504868 - 6924071 on Build GRCh38 | Duplication | MIR4659A MCPH1 XKR5 AGPAT5 DEFB1 MIR8055 GS1-24F4.2 MIR4659B MCPH1-AS1 ANGPT2 |
nsv831220 | Chr.8:6670324 - 6870969 on Build GRCh38 | Gain | MIR4659A XKR5 AGPAT5 DEFB1 GS1-24F4.2 MIR4659B MCPH1-AS1 |
nsv610006 | Chr.8:6710680 - 7063882 on Build GRCh38 | Gain | DEFT1P2 MIR4659A DEFA3 DEFA6 AGPAT5 DEFB1 DEFT1P DEFA5 GS1-24F4.2 MIR4659B DEFA8P DEFA1 XKR5 DEFA10P DEFA4 DEFA7P DEFA11P DEFA1B DEFA9P |
nsv1031577 | Chr.8:6712833 - 8008628 on Build GRCh38 | Gain | DEFB105B USP17L4 DEFA5 MIR4659B ZNF705G DEFB107B SPAG11B DEFA10P DEFA4 LINC00965 DEFT1P2 DEFA3 PRR23D1 AGPAT5 DEFT1P DEFB108P1 GS1-24F4.2 DEFA8P DEFB106A LOC101929400 USP17L3 SPAG11A DEFB104A USP17L1 DEFA9P OR7E125P LOC101928095 MIR4659A DEFA6 DEFB104B FAM66B FAM90A7P DEFB4A DEFB106B DEFB103A XKR5 OR7E154P DEFB105A LOC107986910 FAM90A10P USP17L8 DEFB107A DEFA11P ZNF705B DEFB1 DEFB109P1B DEFB4B DEFA1 PRR23D2 DEFB103B DEFA7P DEFA1B FAM66E |
dgv6876n100 | Chr.8:6687977 - 6812428 on Build GRCh38 | Loss | MIR4659A XKR5 AGPAT5 MIR4659B MCPH1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |