Assay Details
Target Gene Details
Entrez Gene ID: | 23246 |
Gene Name: | block of proliferation 1 |
Gene Aliases: |
- |
Location: |
Chr.8:144262046-144291438 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BOP1 | NM_015201.4 | NP_056016.1 | ||
AB060694.1 | BAB70666.1 | |||
AK024840.1 | ||||
AK290677.1 | ||||
AK293719.1 | ||||
AK315612.1 | ||||
BC001086.2 | AAH01086.2 | |||
BC005160.2 | AAH05160.2 | |||
BC007274.1 | AAH07274.1 | |||
BC013787.1 | AAH13787.1 | |||
BC013980.2 | AAH13980.1 | |||
BC017674.2 | AAH17674.1 | |||
BQ711345.1 | ||||
D50914.1 | BAA09473.1 | |||
DA510177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv12434n54 | Chr.8:144270080 - 144516801 on Build GRCh38 | Loss | TONSL-AS1 MIR6849 SLC52A2 FOXH1 HSF1 MIR1234 MFSD3 BOP1 SCRT1 MIR6848 PPP1R16A FBXL6 MIR6893 CPSF1 LOC101928902 DGAT1 VPS28 KIFC2 RECQL4 TONSL TMEM249 CYHR1 GPT SLC39A4 MIR939 ADCK5 LOC101928953 |
nsv470249 | Chr.8:143889098 - 144519234 on Build GRCh38 | Loss | MIR661 TONSL-AS1 MIR6849 WDR97 SLC52A2 OPLAH FOXH1 HSF1 MIR1234 MFSD3 BOP1 SCRT1 LRRC14 CYC1 MIR6848 PPP1R16A FBXL6 GRINA MIR6847 MIR6893 CPSF1 LOC101928902 DGAT1 VPS28 PLEC SMPD5 HGH1 KIFC2 MROH1 RECQL4 SCX SPATC1 TONSL TMEM249 CYHR1 MIR6846 MAF1 GPT MIR7112 SLC39A4 GPAA1 MIR939 PARP10 ADCK5 EXOSC4 LOC101928953 SHARPIN |
nsv515809 | Chr.8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | MIR661 TONSL-AS1 MIR6849 WDR97 SLC52A2 OPLAH FOXH1 HSF1 MIR1234 MFSD3 BOP1 SCRT1 LRRC14 CYC1 MIR6848 PPP1R16A FBXL6 GRINA MIR6847 MIR6893 CPSF1 LOC101928902 DGAT1 VPS28 PLEC SMPD5 HGH1 KIFC2 MROH1 RECQL4 SCX SPATC1 TONSL TMEM249 CYHR1 MIR6846 MAF1 GPT MIR7112 SLC39A4 GPAA1 MIR939 PARP10 ADCK5 EXOSC4 LOC101928953 SHARPIN |
nsv1025071 | Chr.8:144044224 - 144300847 on Build GRCh38 | Gain | SMPD5 HGH1 MROH1 WDR97 OPLAH HSF1 SCX SPATC1 BOP1 CYC1 MIR6846 MAF1 MIR7112 MIR6847 GPAA1 EXOSC4 SHARPIN |
nsv428208 | Chr.8:144041826 - 144722291 on Build GRCh38 | Gain | TONSL-AS1 MIR6849 WDR97 SLC52A2 OPLAH FOXH1 HSF1 MIR1234 MFSD3 LRRC24 BOP1 SCRT1 LRRC14 CYC1 MIR6848 PPP1R16A FBXL6 ZNF251 MIR6847 MIR6893 CPSF1 LOC101928902 DGAT1 VPS28 SMPD5 HGH1 KIFC2 MROH1 RECQL4 SCX SPATC1 TONSL TMEM249 CYHR1 MIR6846 MAF1 GPT MIR7112 ARHGAP39 SLC39A4 GPAA1 MIR939 ADCK5 EXOSC4 LOC101928953 C8orf82 SHARPIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |