Assay Details
Target Gene Details
Entrez Gene ID: | 761 |
Gene Name: | carbonic anhydrase 3 |
Gene Aliases: |
CAIII, Car3 |
Location: |
Chr.8:85438827-85449040 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CA3 | NM_005181.3 | NP_005172.1 | ||
AK096880.1 | ||||
AK300670.1 | ||||
AK300677.1 | ||||
AK313254.1 | ||||
BC004897.2 | AAH04897.1 | |||
DA421884.1 | ||||
EU794596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831377 | Chr.8:85350878 - 85531912 on Build GRCh38 | Gain | CA3 CA1 CA3-AS1 CA2 |
nsv1028148 | Chr.8:85398341 - 85640901 on Build GRCh38 | Gain | CA3 CA3-AS1 CA2 |
esv2761206 | Chr.8:85398353 - 85556639 on Build GRCh38 | Gain | CA3 CA3-AS1 CA2 |
nsv483126 | Chr.8:83687766 - 85887771 on Build GRCh38 | Gain+Loss | CA13 REXO1L1P LOC101929627 LRRCC1 C8orf59 LOC100288562 RALYL LOC102723322 LOC101929601 CA3-AS1 E2F5 CA2 LOC107986879 CA3 REXO1L2P CA1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |