Assay Details
Target Gene Details
Entrez Gene ID: | 142685 |
Gene Name: | ankyrin repeat and SOCS box containing 15 |
Gene Aliases: |
- |
Location: |
Chr.7:123567003-123637900 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASB15 | XM_011515819.2 | XP_011514121.1 | ||
XM_017011757.1 | XP_016867246.1 |
Target Gene Details
Entrez Gene ID: | 4698 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A5 |
Gene Aliases: |
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13 |
Location: |
Chr.7:123536997-123601651 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA5 | NM_001291304.1 | NP_001278233.1 | ||
NR_111926.1 | ||||
BF790429.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831116 | Chr.7:123447558 - 123619922 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
nsv524192 | Chr.7:122763367 - 123578468 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv508479 | Chr.7:123551811 - 123612091 on Build GRCh38 | Deletion |
![]() ![]() |
esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)