Assay Details
Target Gene Details
Entrez Gene ID: | 57224 |
Gene Name: | NHS like 1 |
Gene Aliases: |
C6orf63 |
Location: |
Chr.6:138422043-138692616 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NHSL1 | NM_001144060.1 | 6 | 993 | NP_001137532.1 |
NM_020464.1 | 5 | 1473 | NP_065197.1 | |
XM_005267062.4 | 6 | 3257 | XP_005267119.1 | |
XM_011535965.2 | 7 | 3311 | XP_011534267.1 | |
XM_011535966.2 | 8 | 2987 | XP_011534268.1 | |
XM_011535967.2 | 8 | 2931 | XP_011534269.1 | |
XM_011535968.2 | 6 | 3179 | XP_011534270.1 | |
XM_011535969.2 | 7 | 2637 | XP_011534271.1 | |
XM_011535970.2 | 7 | 3501 | XP_011534272.1 | |
XM_011535971.2 | 6 | 1768 | XP_011534273.1 | |
XM_011535972.2 | 7 | 1582 | XP_011534274.1 | |
XM_011535973.2 | 6 | 1934 | XP_011534275.1 | |
XM_011535974.2 | 7 | 2726 | XP_011534276.1 | |
XM_011535976.1 | 7 | 978 | XP_011534278.1 | |
XM_017011086.1 | 8 | 2897 | XP_016866575.1 | |
XM_017011087.1 | 7 | 1931 | XP_016866576.1 | |
XM_017011088.1 | 5 | 2525 | XP_016866577.1 | |
XM_017011089.1 | 6 | 1526 | XP_016866578.1 | |
AK307584.1 | 6 | 993 | ||
BX647246.1 | 2 | 366 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2752090 | Chr.6:138395100 - 138705640 on Build GRCh38 | Gain | MIR3145 FLJ46906 HEBP2 NHSL1 |