Assay Details
Target Gene Details
Entrez Gene ID: | 441094 |
Gene Name: | NR2F1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:93409356-93581320 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NR2F1-AS1 | NR_021491.2 | |||
NR_109820.1 | ||||
NR_109821.1 | ||||
NR_109822.1 | ||||
NR_109823.1 | ||||
NR_109824.1 | ||||
NR_109825.1 | ||||
AK124699.1 | ||||
AK296281.1 | ||||
AK311143.1 | ||||
BX647510.1 | ||||
DB257894.1 | ||||
HY108556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830402 | Chr.5:93368195 - 93511639 on Build GRCh38 | Gain | NR2F1-AS1 |