Assay Details
Target Gene Details
Entrez Gene ID: | 55247 |
Gene Name: | nei like DNA glycosylase 3 |
Gene Aliases: |
FGP2, FPG2, NEI3, ZGRF3, hFPG2, hNEI3 |
Location: |
Chr.4:177309837-177362938 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEIL3 | NM_018248.2 | NP_060718.2 | ||
XM_017008360.1 | XP_016863849.1 | |||
AB079071.1 | BAC06479.1 | |||
AK001720.1 | BAA91860.1 | |||
AK307692.1 | ||||
BC025954.1 | AAH25954.1 | |||
BE865481.1 | ||||
BP380910.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3603300 | Chr.4:177297546 - 177343289 on Build GRCh38 | Gain |
![]() |
esv2759304 | Chr.4:177244315 - 177484220 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3894076 | Chr.4:177303484 - 177340740 on Build GRCh38 | Loss |
![]() |
esv34846 | Chr.4:177307859 - 177379956 on Build GRCh38 | Gain |
![]() |
nsv822852 | Chr.4:177295992 - 177380771 on Build GRCh38 | Gain |
![]() |
esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)