Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAND2-AS1 | NR_003679.2 | |||
NR_136192.1 | ||||
NR_136193.1 | ||||
NR_136194.1 | ||||
NR_136195.1 | ||||
NR_136196.1 | ||||
NR_136197.1 | ||||
NR_136198.1 | ||||
NR_136199.1 | ||||
NR_136200.1 | ||||
NR_136201.1 | ||||
AB075501.1 | BAE45751.1 | |||
AK054605.1 | ||||
AK123668.1 | ||||
AK300846.1 | ||||
AU253586.1 | ||||
BG519698.1 | ||||
BQ882603.1 | ||||
DA665500.1 | ||||
DA863153.1 | ||||
DB292439.1 | ||||
DC386781.1 | ||||
DC421261.1 | ||||
EF529495.1 | ||||
EF529496.1 | ||||
EF529497.1 | ||||
EF529498.1 | ||||
EF529499.1 | ||||
EF529500.1 | ||||
EF529501.1 | ||||
EF529502.1 | ||||
EF529503.1 | ||||
EF529504.1 | ||||
EF529505.1 | ||||
EF529506.1 | ||||
EF529507.1 | ||||
EF529508.1 | ||||
EF529509.1 | ||||
EU000853.1 | ||||
EU000854.1 | ||||
EU000855.1 | ||||
EU000856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522261 | Chr.4:173522345 - 173538672 on Build GRCh38 | Loss | HAND2-AS1 HAND2 |