Assay Details
Target Gene Details
Entrez Gene ID: | 9797 |
Gene Name: | TatD DNase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.3:10248493-10281222 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TATDN2 | NM_014760.3 | NP_055575.3 | ||
AK289794.1 | ||||
BC090935.1 | AAH90935.1 | |||
BC101770.1 | AAI01771.1 | |||
BC101776.1 | AAI01777.1 | |||
D86972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv436362 | Chr.3:10048344 - 11860564 on Build GRCh38 | Deletion | SEC13 SLC6A1 GHRLOS LOC101927467 HRH1 SLC6A1-AS1 FANCD2 IRAK2 ATP2B2 LINC00852 GHRL FANCD2OS VHL TAMM41 LINC00606 ATP2B2-IT2 BRK1 ATG7 SLC6A11 MIR378B MIR885 TATDN2 VGLL4 |
nsv589650 | Chr.3:10260488 - 10294102 on Build GRCh38 | Loss | GHRLOS TATDN2 LINC00852 GHRL |
dgv762e201 | Chr.3:10008967 - 11913136 on Build GRCh38 | Deletion | SEC13 GHRLOS HRH1 SLC6A1-AS1 ATP2B2 FANCD2OS TAMM41 LINC00606 ATP2B2-IT2 BRK1 ATG7 SLC6A11 MIR378B MIR885 CIDECP EMC3 VGLL4 LOC401052 SLC6A1 LOC101927467 FANCD2 IRAK2 LINC00852 GHRL VHL FANCD2P2 TATDN2 |
nsv1121603 | Chr.3:10071433 - 11889279 on Build GRCh38 | Deletion | SEC13 SLC6A1 GHRLOS LOC101927467 HRH1 SLC6A1-AS1 FANCD2 IRAK2 ATP2B2 LINC00852 GHRL FANCD2OS VHL TAMM41 LINC00606 ATP2B2-IT2 BRK1 ATG7 SLC6A11 FANCD2P2 MIR378B MIR885 TATDN2 VGLL4 |
nsv236904 | Chr.3:10266949 - 10274997 on Build GRCh38 | Deletion | TATDN2 |
nsv436869 | Chr.3:10064456 - 11877985 on Build GRCh38 | Insertion | SEC13 SLC6A1 GHRLOS LOC101927467 HRH1 SLC6A1-AS1 FANCD2 IRAK2 ATP2B2 LINC00852 GHRL FANCD2OS VHL TAMM41 LINC00606 ATP2B2-IT2 BRK1 ATG7 SLC6A11 FANCD2P2 MIR378B MIR885 TATDN2 VGLL4 |
nsv1073204 | Chr.3:9996548 - 11875005 on Build GRCh38 | Deletion | SEC13 GHRLOS HRH1 SLC6A1-AS1 ATP2B2 FANCD2OS TAMM41 LINC00606 ATP2B2-IT2 BRK1 ATG7 SLC6A11 MIR378B MIR885 CIDECP EMC3 VGLL4 LOC401052 SLC6A1 LOC101927467 EMC3-AS1 FANCD2 IRAK2 LINC00852 GHRL VHL FANCD2P2 TATDN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |