Assay Details
Target Gene Details
Entrez Gene ID: | 100874330 |
Gene Name: | long intergenic non-protein coding RNA 1014 |
Gene Aliases: |
KCNMB2-IT1 |
Location: |
Chr.3:178419201-178457305 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01014 | NR_046786.1 | |||
BX105665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525869 | Chr.3:178401581 - 178436727 on Build GRCh38 | Gain | LINC01014 |
nsv513734 | Chr.3:178398576 - 178439042 on Build GRCh38 | Insertion | LINC01014 |
nsv4136 | Chr.3:178386237 - 178431491 on Build GRCh38 | Deletion | LINC01014 |
nsv4137 | Chr.3:178420749 - 178437917 on Build GRCh38 | Insertion | LINC01014 |