Assay Details
Target Gene Details
Entrez Gene ID: | 254827 |
Gene Name: | N-acetylated alpha-linked acidic dipeptidase-like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:174439669-175809510 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Target Gene Details
Entrez Gene ID: | 100862679 |
Gene Name: | NAALADL2 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.3:175079312-175115242 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAALADL2-AS3 | NR_046390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829799 | Chr.3:175000333 - 175179508 on Build GRCh38 | Gain | NAALADL2 NAALADL2-AS3 |
nsv829798 | Chr.3:174915148 - 175091082 on Build GRCh38 | Gain | NAALADL2 NAALADL2-AS3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |