Assay Details
Target Gene Details
Entrez Gene ID: | 5276 |
Gene Name: | serpin family I member 2 |
Gene Aliases: |
MEPI, PANCPIN, PI14, TSA2004 |
Location: |
Chr.3:167441789-167474132 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINI2 | NM_001012303.2 | NP_001012303.1 | ||
NM_006217.4 | NP_006208.1 | |||
AB006423.1 | BAA33766.1 | |||
AF130470.1 | AAD34723.1 | |||
AK226025.1 | ||||
AK293391.1 | ||||
BC027859.2 | AAH27859.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv592495 | Chr.3:167077165 - 167655374 on Build GRCh38 | Gain | SERPINI2 ZBBX WDR49 LINC01327 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |